About this item
The Barnstead Pacific TII water purification systems provides up to 240L/day of Type 2 (10-15 megohm) water from tap water
The Pacific TII systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. The 30L and 60L reservoirs may also be mounted on the wall, on the bench, or under the bench.
The Pacific TII systems are ideal for producing type 2 for the rinsing of glassware, supplying lab equipment such as autoclaves or type 1 systems that require pretreated feedwater, or the preparation and diluting of buffers, reagents and media.
All Pacific TII systems include: quick discconnects to make changing consumables easy, and all core consumables and tubing. Pacific TII systems are offered in five output volumes, 3L/hr, 7L/hr, 12L/hr, 20L/hr and 40L/hr. All models can be ordered with or without a built in UV lamp. External reservoirs are required and are offered in three different sizes, 30L, 60L or 100L.
Specifications
- Catalog No:
- CA89230-396
- CA89230-392
- CA89230-408
- CA89230-406
- CA89230-398
- CA89230-400
- W×D×H:
- 372×330×603 mm (14.6×13×23.7")
- 372×330×603 mm (14.6×13×23.7")
- 372×330×603 mm (14.6×13×23.7")
- 372×330×603 mm (14.6×13×23.7")
- 372×330×603 mm (14.6×13×23.7")
- 372×330×603 mm (14.6×13×23.7")