Specifications
- W×D×H:450×580×1500 mm (17.7×22.8×59.1")
- Description:LabTower RO 40 LPH System, 40 L/hr output
- Production flow rate:40 L/h
- Cat. no.:89230-448
- Supplier No.:50132391
Specifications
About this item
The Barnstead LabTower RO water purification systems provides up to 200L/day of Type 3 (reverse osmosis) water from tap water
The LabTower RO systems are completely free-standing units which require no bench or wall space. The unit comes standard with casters for easy moving.
The LabTower RO systems are suited to meet a wide range of applications needs. Type 3 (RO) water can be used for rinsing glassware, supplying water to autoclaves, waterbaths, and preparing or diluting buffers, reagents or media for example.
All LabTower RO systems include: quick discconnects to make changing consumables easy, built in 100L reservoir and all core consumables and tubing. LabTower RO systems are offered in two output volumes, 20L/hr or 40L/hr. Both models come standard with a 100L integrated reservoir.