SP Bel-Art Easy-Read Individually Calibrated Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Thermometers contain black, non-toxic, biodegradable, EnviroKleen™ certified, Enviro-Safe® liquid against lead-free, yellow back glass for easy readability and fewer reading errors.
- Non-toxic and biodegradable liquid
- Lead-free
- Yellow back glass
- Easy to read
They are ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. Individually serialized and made in the USA.
Ordering information: Each easy-to-read thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.
Specifications
- Catalog No:
- 75795-778
- 30617-782
- 89218-020
- 75795-782
- 75795-784
- 89082-179
- 30617-808
- 89218-024
- 89218-028
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
Specifications
Related Information
AT-VWR Thermometer FAQs
Datasheet
Bel-Art_New_Product_Supplement_Laboratory_Problem_Solvers_Brochure
Product Literature
ED-EnviroKleenCertification
Additional Product Details
ghs_B60202