DuPont™ Tychem® 4000 Bib Aprons
Catalog #: CA13452-842
Supplier: DuPont
Specifications
- Garment application:Industrial,Paint and lacquer,Chemical protection
- Length:431/2"
- Color:White
- Height:273/4"
- Cat. no.:CA13452-842
- Supplier No.:SL274BWH0000
Specifications
About this item
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
- Neck loop
- Waist ties
- Bound seams
- One size fits most
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
These bib aprons provide enhanced frontal protection. Bound seams and ties at waist allow for one size to fit most.
Certifications: Thickness meets ASTM D1777.
Customers Who Bought This Also Bought