About this item
Tychem® RESPONDER® CSM combines multiple film barriers laminated to both sides of a high-strength polypropylene substrate for high level protection against toxic and corrosive chemicals.
- Expanded back
- Front entry
- Standard visor
- Attached butyl gloves
- Attached socks with outer boot flaps
- Double storm flap with hook and loop closure
- Two exhaust valves
- Double-taped seams
It is effective for handling chemical warfare agents as well as military site cleanup and HazMat first response. Tychem® RESPONDER® CSM fabric provides at least 30 minutes of protection against 320 chemical challenges. Each suit undergoes rigorous inspection and each Level A suit, pressure testing. Each fabric lot is tested using live toxic agents plus one garment for each test fabric production run is also tested for seam barrier with live toxic agents. Coveralls are a desert tan color for low visibility applications.
The encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment. Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing. Faceshields are triple-layered (PVC 40 mil/Teflon® 5 mil/PVC 20 mil) and standard width. Front entry design allows wearer to partially don garment (maintain standby position) without assistance. Expanded back accommodates self-contained air breathing apparatus (SCBA). Coveralls feature a vapor tight zipper, which is extra long to increase garment opening and aid donning and doffing. Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion. Two exhaust valves release higher pressure inside encapsulating garments, while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back. Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
Specifications
- Catalog No:
- CA13452-712
- CA13452-714
- CA13452-708
- CA13452-710
- CA13452-716
- CA13452-718
- Garment application:
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
Specifications
Related Information
Doffing Level A and Level B DuPont™ Tychem® Encapsulated Suits Doffing Level A and Level B DuPont™ Tychem® Encapsulated Suits Field Example Donning Level A & Level B DuPont™ Tychem® Encapsulated Suits Encapsulated Suit Pressure Test Folding An Encapsulated Suit
Videos
VWR® Focus Production Volume 3