Zinc Oxide
Supplier: Ward's Science
Certificates
About this item
CAS Number: 1314-13-2
Formula Weight: 81.39
Formula: ZnO
Hazard Info: Toxic (fumes)
Density (g/mL): 5.1
Freezing Point (°C): 1975
Solubility: Acids and Alkalies
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together