To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Wrinkler's Solution #2
- Size:500 mL
- Chemical/Physical Specifications:#2
- Cat. no.:470303-046
- Grade:Reagent
- Supplier No.:WW0030-500MLR
- Disposal Method:This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.Use a lab apron, chemical resistant gloves and goggles before following the below method to dispose of hazardous waste.1. The concentration of the base should be 1 M or less. Dilute it to this concentration if it is more concentrated. Use a large beaker and place it in a fume hood.a. When dissolving a base, heat may be evolved. Ice can be used to help control the heat. b. Work with a small amount of base at first, and then scale up.2. Slowly add dilute hydrochloric acid to the base until the mixture is neutralized to about a pH of 7.3. Pour the final solution down the drain with at least a 20-fold excess of water.
- Solution Preparation:Mix 500 g of sodium hydroxide and 135 g of sodium iodide in 600 mL of DI water. Place beaker in an ice bath. Dilute to a final volume of 1 L with DI water.
Specifications
About this item
Synonyms: Alkaline-Iodide
Shelf Life (months): 36
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.