What do Seababies tell us about Pollution Lab Activity
Catalog #: 470024-486
Supplier: EDVOTEK
Specifications
- Description:What do Seababies tell us about Pollution Lab Activity
- No. of Lab Groups:6
- Cat. no.:470024-486
- Supplier No.:DD-304
Specifications
About this item
Investigate Pollution
- Simulates environmental incident
- Open or guided inquiry
Demonstrate how pollutants have deleterious consequences on local seawater ecology. In this experiment, the brine shrimp, Artemia franciscana, are hatched from eggs in artificial seawater and the effect of various simulation toxins are added. The effect of simulated toxicants will be monitored by their impact on growth and survival.
Customers Who Bought This Also Bought