Specifications
- Description:WaterWorks Bacteria Check
- No. of Lab Groups:1
- Cat. no.:470106-166
- Supplier No.:487197
Specifications
About this item
Bacteria Make Up Much of the Composition of the Soil
- Aligned to USEPA standards
- Equipment is sterile
- Includes instructions
A test bottle for the rapid detection of coliforms in water. The pre-measured media makes this test ideal for field, on-site or lab use.
WaterWorks Bacteria Check contains sterile Presence-Absence media. Includes very simple, step-by-step instructions to help you detect if any harmful bacteria (coliform) are present in your water supply. Use Bacteria Check as a classroom demonstration or in your home to test your own water supply. The test requires that the specimen be undisturbed for 48 hours.
USEPA Compliant, Sterile