VWR® Pipette Fillers
Catalog #: 470123-222
Supplier: Heathrow Scientific
Specifications
- Accuracy:±0.01 ml
- Cat. no.:470123-222
- Supplier No.:HS20632A
Specifications
About this item
Pipette fillers eliminate the hazardous practice of drawing liquids into pipettes by mouth.
- Filters work with pipettes up to 50 ml in size
- Filters help avoid sample contamination
- Safe, precise delivery
- Ideal for volatile, corrosive, or poisonous liquids
- Fits a standard serological pipet
To use, insert the pipette into the bottom opening of the filler, draw to the desired volume, and expel by pressing lettered valves.
The filters are made of soft rubber. Colors may vary. Dimensions: 54×54×130 mm (L×W×D).
Customers Who Bought This Also Bought