Volumetric Transfer Pipettes
Catalog #: 470016-708
Supplier: Avantor
Specifications
- Description:Volumetric transfer pipettes, thin stem, overall length: 15.0 cm, stem diameter: 2.5 mm, bulb draw: 3.1 ml, capacity: 4.0 ml, drops per ml: 250, pack of 100 pipettes
- Cat. no.:470016-708
- Supplier No.:WARD470016-708
Specifications
About this item
Economical and convenient polyethylene transfer pipettes are chemically inert to most biologicals and acids. Designed for consistent drop sizes, making these pipettes reliable in many experiments where liquids are transferred. Non-sterile.
Customers Who Bought This Also Bought