Specifications
- Description:Urinary Blood Test Strips, Vial of 100
- Cat. no.:470019-032
- Supplier No.:URS-01PR-BL77
Specifications
About this item
Includes Color Key for Easy Reading of Results
Simply place a drop of urine on the strip’s indicator spot and wait one minute for the color change. These strips are clinically tested for accuracy and have a shelf life of two years if kept in the resealable dry containers. They can also be used with WARD’S Artificial Urine. Vials of 100.