Tray/Tote Box
Catalog #: 470173-908
Supplier: Dynalon
Specifications
- Description:Tray/Tote Box
- Cat. no.:470173-908
- Supplier No.:419244
Specifications
About this item
Designed to nest together without sticking, this tray is molded of hefty linear polyethylene. It will not corrode or mildew, and it remains chemically resistant to alkalis, oils, acids, and detergents. Size: 21"L x 12"W x 6"D.
Customers Who Bought This Also Bought