Toxoplasma gondii, Small Intestine Slide
Catalog #: 470181-970
Supplier: Avantor
Specifications
- Description:Toxoplasma gondii , Small Intestine
- Cat. no.:470181-970
- Stain:Iron Hematoxylin
- Supplier No.:WARD470181-970
- Orientation:Section
Specifications
About this item
A Common Parasite in Cats
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows schizogony and gametogony.
Customers Who Bought This Also Bought