Touch Test Two-Point Discriminator
Catalog #: 470019-284
Supplier: Fabrication Enterprises
Specifications
- Description:Two-Point Discriminator
- Size:2½" dia.
- Cat. no.:470019-284
- Supplier No.:12-1495
Specifications
About this item
Test Static And Moving Discrimination
Two sturdy plastic wheels joined together have tips spaced at standard testing intervals. The unit comes with instructions and a protective carrying case.
Customers Who Bought This Also Bought