Specifications
- Description:TI-84 Plus EZ Spot teacher kit
- Cat. no.:CA-470350-662
Specifications
About this item
Calculate graphical equations with accuracy and speed.
The TI-84 Plus features 480 KB FLASH ROM memory for data archive and storage of Apps, 24KB of RAM, USB port for computer connectivity, and 8-line by 16-character display. Real and complex numbers can be calculated to 14-digit accuracy and displayed with 10 digits plus a 2-digit exponent. Graphs 10 rectangular functions, 6 parametric expressions, 6 polar expressions, and 3 recursively-defined sequences.Allows for interactive analysis of function values, roots, maximums, minimums, integrals, and derivatives. Matrix operations including inverse, determinant, transpose, augment, reduced row echelon form, and elementary row operations. Convert matrices to lists and vice-versa. Provides 7 different graph styles for differentiating the look of each graph drawn.Business functions including Time-Value-of-Money (TVM), cash flows, and amortization. Full screen interactive editor for solving TVM problems.Powered by 4 AAA batteries with a Silver Oxide battery backup to protect RAM memory during main battery change. Comes with Impact-resistant slide case. One-year limited warranty.
The Silver Edition has the same functionality as the TI-84 Plus, but has been upgraded with 1.5 MB of FLASH ROM memory and a kick-stand slide case. Changeable faceplates are also available for the Silver Edition.
The School Pack of 10 enables teachers to keep a close eye on calculators with brightly-colored yellow back panels. Package includes five USB cables, five unit-to-unit cables, 40 AAA batteries, product CD, user manual, poster, and transparency.
- Ideal for Pre-Algebra, Algebra 1 and 2, Geometry, Statistics, Biology, Physics, Business and Finance
- Permitted for use on SAT, PSAT/NMSQT, ACT, AP, IB and Praxis exams
- Upgradeable operating system
- Statistics functions, including hypothesis testing and calculation of confidence intervals
- Probability distributions
The TI-84 Plus features 480 KB FLASH ROM memory for data archive and storage of Apps, 24KB of RAM, USB port for computer connectivity, and 8-line by 16-character display. Real and complex numbers can be calculated to 14-digit accuracy and displayed with 10 digits plus a 2-digit exponent. Graphs 10 rectangular functions, 6 parametric expressions, 6 polar expressions, and 3 recursively-defined sequences. Allows for interactive analysis of function values, roots, maximums, minimums, integrals, and derivatives. Matrix operations including inverse, determinant, transpose, augment, reduced row echelon form, and elementary row operations. Convert matrices to lists and vice-versa. Provides 7 different graph styles for differentiating the look of each graph drawn. Business functions including Time-Value-of-Money (TVM), cash flows, and amortization. Full screen interactive editor for solving TVM problems. Powered by 4 AAA batteries with a Silver Oxide battery backup to protect RAM memory during main battery change. Comes with Impact-resistant slide case. 1-year limited warranty.