Thermometer Only
Catalog #: 470110-970
Supplier: Peco Sales
Specifications
- Description:Thermometer Only, Permacolor-Filled Thermometer
- Cat. no.:470110-970
- Supplier No.:89193-C
Specifications
About this item
Permacolor-filled thermometer features projection at top to fit positioning hole.
For replacement of either wet or dry bulb thermometer in Sling Psychrometer. Permacolor-filled thermometer features projection at top to fit positioning hole.
Customers Who Bought This Also Bought