Ward's® Tenacity Collection
Catalog #: 470025-246
Supplier: Avantor
Specifications
- Description:Tenacity Collection
- Cat. no.:470025-246
- Supplier No.:WARD470025-246
Specifications
About this item
Five Types of Cohesiveness
Different types of tenacity are shown in five popular minerals. The set is housed in a compartmented collection box, and an identification list is included.Contents:5 numbered samples, 1" x 1"Minerals:Brittle: sulfurSectile: seleniteMalleable: copperFlexible: vermiculiteElastic: mica
Customers Who Bought This Also Bought