TeacherGeek Clips Pack of 100
Catalog #: 470132-482
Supplier: TEACHERGEEK INC
Specifications
- Description:TeacherGeek Clips Pack of 100
- Cat. no.:470132-482
- Supplier No.:27710
- No. of Parts:100
Specifications
About this item
Bulk TeacherGeek clips to expand your TeacherGeek collection
TeacherGeek Clips are an innovative way to prevent dowels from sliding out of position and to maintain pulley and gear alignment. They are clipped into place by pressing over the contour of the dowel. After aligning gears, pulleys and wheels, the clips are simply snapped into place.
TeacherGeek Clips are an innovative way to prevent dowels from sliding out of position and to maintain pulley and gear alignment. They are clipped into place by pressing over the contour of the dowel. After aligning gears, pulleys and wheels, the clips are simply snapped into place.
Customers Who Bought This Also Bought