Student Plasticware Set
Catalog #: 470039-358
Supplier: Shiv Dial Sud & Sons
Specifications
- Description:Student Plasticware Set
- Cat. no.:470039-358
- Supplier No.:PLKIT20
Specifications
About this item
The assortment of labware contains some of the most commonly used pieces. The plastic set comes with 20 items packed in a plastic storage box.
Delivery: The set includes 2 Beakers (100 and 250 ml), 2 Graduated cylinders (10 and 100 ml), 1 Funnel, 6 mm top dia., 1 drop dispenser bottle, 3 test tubes, 4 reagent bottles, 1 wash bottle, 6 Pipets.
Customers Who Bought This Also Bought