Streak and Color Collection
Catalog #: 470016-522
Supplier: Avantor
Specifications
- Description:Streak and Color Collection
- No. of Samples:9
- Cat. no.:470016-522
- Supplier No.:WARD470016-522
Specifications
About this item
- Illustrates difference between streak and color
- Includes a streak plate
The powder a mineral produces (streak) is sometimes a different color than the mineral first appears. The 9 samples in this set were carefully chosen to help clearly demonstrate this physical property. A streak plate is also included.
Customers Who Bought This Also Bought