Straight Filament Lamp Bulbs
Supplier: GE
Certificates
About this item
Single-line filament is ideal for student diffraction experiments. It's also a great way for students to observe filament technology in action, because the slender 15.2 cm lamp lets them clearly and distinctly see the entire process.
Medium screw base and 120 V, suitable for standard sockets.
Customers Who Bought This Also Bought