Straight Connecting Tubes
Catalog #: 470174-240
Supplier: Dynalon
Specifications
- Description:Tube, Connecting, Straight, 4-6 mm OD
- Cat. no.:470174-240
- Supplier No.:227235
Specifications
About this item
These autoclavable straight connectors are molded of polypropylene and have serrations to hold tubing tightly and prevent slipping. They have excellent chemical and temperature resistance.
Customers Who Bought This Also Bought