Specifications
- Description:Solar Hydrogen Education Set
- No. of Lab Groups:1
- Cat. no.:470039-904
- Supplier No.:FCJJ-16
Specifications
About this item
Introduce the Value of Reducing Carbon Footprint.
Your students may understand the importance of being conscious to the environment but have they ever explored alternative energy solutions to reduce their carbon footprint? This set enables your students the ability to explore their creative side as it provides them the flexibility of inventing their own clean energy applications to generate solar energy and water through the use of fuel cells and renewable hydrogen. Students may also use the included small electric motor and propeller blade as a launching point for motorized applications. This set comes with a complete curriculum on renewable energy with an assembly guide, flash animations, background history on the technology, and an experiment manual that is easy to follow and comprehend.
This science kit should only be used by persons 12 years old and up, and only under the supervision of adults who have familiarized themselves with the safety measures described in the science kit. Keep small children and animals away, as it contains small parts that could be swallowed. The fuel cell generates gases that are very easily ignited. Read the instructions before use and have them ready for reference.
- Design various applications of solar energy
- Age 12 and Up
- Includes instructions
The Solar Hydrogen Science Kit lets students invent their own clean energy applications using fuel cells and renewable hydrogen created using solar energy and water. The kit also comes with a small electric motor and propeller blade as the starting point for motorized applications you can build using your futuristic solar energy storage device. The set comes with a complete curriculum on renewable energy with easy to follow experiment manual, assembly guide, flash animations, and background history on the technology.
* Includes Horizon's complete curriculum on renewable energy
* Lessons on solar energy, hydrogen fuel cells, electrolysis, and basic electronics
This science kit should only be used by persons 12 years old and up, and only under the supervision of adults who have familiarized themselves with the safety measures described in the science kit. Keep small children and animals away, as it contains small parts that could be swallowed. The fuel cell generates gases that are very easily ignited. Read the instructions before use and have them ready for reference.