ContactUsLinkComponent
Ward's® Soils and Isopach Mapping Lab Activity
Ward's® Soils and Isopach Mapping Lab Activity
Catalog #: 470023-770
Supplier:  Avantor
WARD'S SOILS & ISOPACH MAPPING LAB
Ward's® Soils and Isopach Mapping Lab Activity
Catalog #: 470023-770
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Soils and Isopach Mapping Lab Activity
  • Cat. no.:
    470023-770
  • Supplier No.:
    WARD470023-770
Specifications
About this item
Use Real Soil Samples to Analyze Soil Formation and Classify Soil
  • Materials for 30 students
  • Teach students about the formation of soils
Take students on a field trip without leaving the classroom! Students get practical geological field experience by studying and analyzing data and samples from the Wood River Quadrangle in Illinois and Missouri. Students also learn to think like geologists, applying the field evidence gathered to make decisions regarding economic and environmental land use. The lab activity comes with all the materials you need to outfit a class of thirty students.
Students test, evaluate, and classify actual soil samples from the Wood River quadrangle to learn about soil types and how soil is formed. They also construct an isopach map using loess thickness information, and use their results to address questions about land use applications.
Ordering information: Kit includes eight natural soil samples, sediment comparator cards, one Wood River quadrangle map, acid bottles, testing trays, copymasters for student activities, and a teacher’s guide. Additional copies of the topographic map may be purchased separately.
Customers Who Bought This Also Bought