Ward's® Sodalite
Catalog #: 470015-448
Supplier: Avantor
Specifications
- Description:Sodalite, Student Specimens
- Specimen Dimensions:1×1" - 1×2"
- Supplier No.:WARD470015-448
Specifications
About this item
Deep blue, massive, with nepheline; key feldspathoid.
WARD'S Offers:
Student Specimens. Ideal for students to use for study and identification. Specimens range in size from 1" x 1" to 1" x 2" and come in packages of ten.
Customers Who Bought This Also Bought