Slab Easels
Supplier: JULE-ART INC.
Certificates
About this item
Safely prop up flat specimens such as agate or fossils in a thin matrix. The smaller easel supports slabs up to 1/4" thick, the larger easel easily accommodates thicker slabs and crystal plates.
Slab Easel Size: 3"L x 2 1/2"W x 2 1/4"H. Package of four.
Thick Slab Easel Size: 2 1/2"L x 2 1/4"W x 2 3/4"H. Package of four.
Customers Who Bought This Also Bought