Size Exclusion Chromatography Activity Kit
Catalog #: 470149-898
Supplier: G-Biosciences
Specifications
- Description:Size exclusion chromatography activity kit
- No. of Lab Groups:6 groups of 5
- Cat. no.:470149-898
- Supplier No.:BE-414
Specifications
About this item
Demonstrate Principles and Factors of Gel Filtration
This hands-on chromatography lab activity teaches gel filtration or size exclusion chromatography and the use of this method in purification of biological samples. Materials supplied for six student groups of up to five students each.
This hands-on chromatography lab activity teaches gel filtration or size exclusion chromatography and the use of this method in purification of biological samples. Materials supplied for six student groups of up to five students each.
Customers Who Bought This Also Bought