ContactUsLinkComponent
Single Action Stopwatch
Single Action Stopwatch
Catalog #: 470018-448
STOPWATCH SINGLE ACTION 1/5 SEC 30 MIN
Single Action Stopwatch
Catalog #: 470018-448
Specifications
  • Description:
    Stopwatch, single action, 1/5 second, 30 minutes
  • Cat. no.:
    470018-448
  • Supplier No.:
    211003-B
Specifications
About this item
An excellent heavy-duty stopwatch that is ideal for student use.
  • The stopwatch can be read to ¹/₅ second and registers to 30 minutes
  • Attractive metal case
Operation is simple with start, stop, and reset controlled by successive crown depressions.
Customers Who Bought This Also Bought