Specifications
- Description:Shampoo Evaluation Lab Activity
- No. of Lab Groups:12
- Cat. no.:470007-766
- Includes:Teacher's Guide, Student Data Sheets, Assessment Manuals
- Supplier No.:IS9353
Specifications
About this item
Students use science to test their favorite shampoos.
- Students Research the Effects of Everyday Products
- Tests for Relative Viscosity, pH, Oil Emulsification, and More
- Real-World Application
Different shampoos are manufactured from a variety of ingredients to perform a variety of tasks. In this investigation, students not only learn about the role of several shampoo ingredients but also test several properties of different shampoos, which are included in the kit.
Students determine the relative viscosity, pH, flash-foam formation and retention, oil emulsification, and contaminant-dispersion abilities of different shampoos.