Low-Form Evaporating Dish
Supplier: SILICO & CHEMICO PORCELAIN SE
Certificates
About this item
High-Quality Labware, Low Prices
Our laboratory porcelain is high-quality, yet affordably priced. This low-form evaporating dish has a pourout. It is glazed entirely on the inside and over the upper section of the outside. Note: Fragile, use care when handling.
Our laboratory porcelain is high-quality, yet affordably priced. This low-form evaporating dish has a pourout. It is glazed entirely on the inside and over the upper section of the outside. Note: Fragile, use care when handling.
Customers Who Bought This Also Bought