Screen Set
Catalog #: 470016-298
Supplier: Able Printing Service
Specifications
- Description:Screen Set
- Cat. no.:470016-298
- Supplier No.:3612
Specifications
About this item
Fine heavy-duty Bristol board screens that fit into our screen support on your meter-stick optical bench.
Each one measures 10×12.5 cm and has a millimeter scale along one side.
Customers Who Bought This Also Bought