Sclerenchyma Slide
Catalog #: 470182-528
Supplier: Avantor
Specifications
- Description:Sclerenchyma
- Cat. no.:470182-528
- Stain:Quadruple stained
- Supplier No.:WARD470182-528
- Orientation:Cross section
Specifications
About this item
Heavily Stabilized Cell Walls
- Identifying features clearly distinguishable
- Rigorous quality control standards
Aristolochia. Provide structural support in plants.
Customers Who Bought This Also Bought