Specifications
- Description:Ward's® Chemical Battery Demonstration
- Cat. no.:470010-848
- Includes:Instructions
- Supplier No.:WARD470010-848
- No. of Uses:5 - 7
Specifications
About this item
Construct a Simple Chemical Wet-Cell Battery.
- The perfect supplement to your chemistry lectures
- Can be completed in 15 minutes
- Requires only basic labware that you provide
This easily-constructed battery can be built using two chemical solutions and two metal electrodes in an Erlenmeyer flask. It produces a constant source of 1.5 V and can be used to power a small LED. Using the materials included in this kit, you can show your students how to construct a single battery of this type.