Specifications
- Description:Robbery At Anytown Mall Lab Activity
- No. of Lab Groups:6
- Cat. no.:470017-714
- Supplier No.:10-548
Specifications
About this item
Practice forensic skills in engaging activity.
- Grades 6-9
- Real world case based scenario
- Combination of 6 activities
Use six different types of trace evidence to solve the crime in this hands-on CSI-like activity. Using the scientific method steps of observation, experimentation, and interpretation, students will analyze hair, fiber, fabric, blood, fingerprints, and handwriting. This evidence coupled with included personal profiles enables students to crack the case. Includes materials for six student groups
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.