Rive Ray Box
Supplier: OSAW INDUSTRIAL PRODUCTS SE
Certificates
About this item
High quality and low cost make this ideally suited for junior high or high school students.
- Includes on/off switch
- Slotted screen for multiple light rays
It is durably constructed with a rigid vinyl base and a metal box housing a transformer and a high intensity light. An adjustable screen produces either one wide beam or a choice of one, three, or five slit beams, all of which can be easily seen tracing a path across the table surface on which the unit sits.
Certifications: CSA approved.
Ordering information: Rive ray box is available as individual or case of 8; slit screen is available as a pack of 5 or case of 50 packs (250 ea.); and the replacement bulb is available as individual or case of 6.
Customers Who Bought This Also Bought