Riester Diagnostic Kit
Catalog #: 470020-422
Supplier: Graham-Field
Specifications
- Description:Riester Diagnostic Kit
- Cat. no.:470020-422
- Supplier No.:1227-4
Specifications
About this item
A rheostat controls the light intensity coming from the incandescent lamps in the instrument heads, and the battery handle utilizes a bayonet for changing the multiple instrument heads in this multipurpose kit.
- Several Instruments for Examining the Eyes, Ears, and Throat
- Bent Arm Illuminator Lights Oral Cavity and Pharyngeal Area
- Nasal Speculum Opens and Closes within a Wide Range
- Rheostat Controls the Light Intensity from Instrument Head Lamps
- Tongue Blade Holder Works with Wooden or Plastic Tongue Depressors
It includes a bent arm illuminator to light the oral cavity and pharyngeal area, two laryngeal mirrors (3 and 4 mm), May ophthalmoscope with lens wheel and correcting lens diopters from -20 to +20, nasal speculum that manually opens and closes within a wide range, and tongue blade holder that can be used with wooden or plastic tongue depressors.
It also includes an otoscope with a removable cap, three reusable specula, swivel magnifying lens, and an outlet for pneumatic testing.
Ordering information: This unit requires two C Batteries, not included.
Packaging: This unit comes packed in a hard storage case.
Customers Who Bought This Also Bought