Specifications
- Description:Refraction Tank with Laser
- Cat. no.:470105-996
- Dimensions:17.1W x 13.9D x 25.4H cm (6.75 x 5.5 x 10")
- Supplier No.:6330-00
Specifications
About this item
The refraction tank with laser provides excellent visibility!
Appropriate for Grades 6-12
The Refraction Tank with Laser provides excellent visibility!
Use this sturdy, self-contained device to measure the refraction and incidence of light as well as the critical angle and total internal reflection. Simply fill the circular acrylic tank halfway with water and use the built-in laser, which can be rotated 360° around the tank, to direct a bright, narrow ray of light onto the water surface. The base contains 4 leveling screws to level the water in the tank.
The unit measures 6.75" W x 5.5" D x 10" H (17.1cm x 13.9cm x 25.4cm) and it requires (1) LR44 button battery (included).
- Rotating laser for easy positioning
- Demonstrate refraction and internal reflection
Use this sturdy, self-contained device to measure the refraction and incidence of light as well as the critical angle and total internal reflection. Simply fill the circular acrylic tank halfway with water and use the built-in laser, which can be rotated 360° around the tank, to direct a bright, narrow ray of light onto the water surface. The base contains 4 leveling screws to level the water in the tank. Appropriate for Grades 6-12. Requires (1) LR44 button battery (included).