Receptacle for Small Bulb
Catalog #: 470111-196
Supplier: United Scientific Supplies
Specifications
- Description:Receptacle for Small Bulb
- Cat. no.:470111-196
- Supplier No.:LMPRO2
Specifications
About this item
This lamp holder is suitable for miniature base lamps.
- Plastic construction
- Easy connect fahnestock clips
This holder includes Fahnestock clips for quick, easy connections.
Customers Who Bought This Also Bought