Specifications
- Description:Pulse Glass
- Cat. no.:470146-174
- Supplier No.:2010-20
Specifications
About this item
Ideal for demonstrating that vapor pressure increases when temperature increases.
Ideal for demonstrating that vapor pressure increases when temperature increases.
Two glass bulbs are connected with a glass tube, and contain a colored, volatile liquid. Holding one of the bulbs in your hand causes the evaporation of a portion of the liquid that forces the liquid to the other bulb. Length is 23.5 cm.
- Easy to use
- Length 23.5 cm
Two glass bulbs are connected with a glass tube, and contain a colored, volatile liquid. Holding one of the bulbs in your hand causes the evaporation of a portion of the liquid that forces the liquid to the other bulb.