Specifications
- Description:PTC Taste Test Paper
- Cat. no.:470123-120
- Supplier No.:165-12V-100
Specifications
About this item
Genetic Test Paper
- Pocket-Sized, Lightweight Vial
- 100 test strips per vial
Teach students about dominant and recessive genes with PTC Taste Test Paper. The ability to taste bitterness of the test paper is usually considered as a dominant genetic trait, whereas the inability to taste the paper is considered a recessive trait. Seven out of ten people are able to taste PTC.