Projectile Launcher Demonstrator
Catalog #: 470014-948
Supplier: Avantor
Specifications
- Description:Projectile Launcher Demonstrator
- Cat. no.:470014-948
- Supplier No.:WARD470014-948
Specifications
About this item
Safe and Convenient Visual Presentation
Consistent, measurable results are achieved every time with this economical demonstration. Students set-up and manipulate the launcher to observe and record the established projectile motion. Illustrate how launch angle, launch velocity, spring tension, and strength all effect projectile motion. For simple demonstrations in the classroom or lab, this device can be set up on any surface using the included C-clamp. Instructions included. Size: 15 cm x 15 cm.
Customers Who Bought This Also Bought