To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Potassium Fluoride
- Size:500 g
- Chemical/Physical Specifications:Powder
- Cat. no.:470302-128
- Grade:Reagent
- Supplier No.:PP0563-500GR
- Disposal Method:This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.
Use a lab apron, chemical resistant gloves and goggles before following the below method to dispose of hazardous waste.
1. For hydrofluoric acid disposal, a plastic container must be used as hydrofluoric acid will react with the glass. The container should be placed in a fume hood.
2. Fill the plastic container with a three-fold molar excess of solid calcium hydroxide, then fill it half full with water.
3. Slowly add the hydrofluoric acid to the calcium hydroxide suspension.
a. Heat may be evolved, if necessary, cool the plastic container in an ice bath.
4. Neutralize the basic solution with dilute hydrochloric acid to a pH of about 7.
a. If the solution becomes too acidic, calcium fluoride will precipitate out.
5. After letting the neutralized suspension sit overnight, decant the liquid.
6. Once the solid has dried, place it in a container and it should be disposed of in a landfill.
7. Pour the final solution down the drain with at least a 20-fold excess of water. - Solution Preparation:To prepare a solution, determine the formula weight of the material, the concentration (Molarity) of the solution, and the amount of solution desired in Liters. Once this information has been obtained, use the following formula:
(Formula Weight) x (Concentration) x (Desired Volume) = Number of grams in desired volume of liquid
For a 1 M, 1 L solution:
(Formula Weight) x (1 mol/L) x (1 L) = x g added to 1-L volumetric flask. Dilute to a final volume of 1 L with DI Water.
Specifications
About this item
CAS Number: 7789-23-3
Formula Weight: 58.10
Formula: KF
Density (g/mL): 2.481
Boiling Point (°C): 1505
Freezing Point (°C): 859
Solubility: Water and Hydrochloric Acid
Shelf Life (months): 36
Storage: Blue
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.