Portable pH Meter
Catalog #: CA39100-600
Supplier: Hanna
Specifications
- Description:Portable pH Meter
- Cat. no.:CA39100-600
- Supplier No.:HI 8424
Specifications
About this item
Housed in a rugged, water-resistant case, this meter is accurate, reliable, and affordable. The standard combination electrode is ideal for general measurements, and the temperature probe has a stainless steel sheath for extra chemical resistance and long life.
- Full 0.00 to 14.00pH Range
- Accurate to ±0.01pH
- Automatic Temperature Compensation
- Push-Button Calibration
Calibration is done with the push of a button—the built-in microprocessor automatically recognizes the buffer being used and calibrates accordingly. The LCD display can show pH, temperature (°C) and mV (for ORP). Powered by a single 9V alkaline battery, the meter accepts any replacement electrode with a BNC connector.
Range: 0.00 to 14.00pH/±400mV/ 0 to 100°C
Resolution: 0.01pH/1mV/0.1°C
Accuracy: ±0.01pH/±1mV/±0.1°C
Dimensions: 7.8" x 3.2" x 1.8"
Customers Who Bought This Also Bought