Ward's® Porphyry
Catalog #: 470015-460
Supplier: Avantor
Specifications
- Description:Porphyry
- Specimen Dimensions:Large hand specimen
- Supplier No.:WARD470015-460
Specifications
About this item
Shows porphyritic texture in a fine dark matrix abundant large feldspar phenocrysts. Hand specimen.
Customers Who Bought This Also Bought