Polypropylene Griffin Graduated Beakers
Catalog #: 470144-856
Supplier: Shiv Dial Sud & Sons
Supplier #: CH10155/1
Log in for availability
About this item
Economical, Unbreakable Beakers.
- Permanent molded graduations
- Special no-drip spouts
- Autoclavable up to 121°C
Customers Who Bought This Also Bought