Polypropylene Buchner Funnel
Catalog #: 470174-670
Supplier: United Scientific Supplies
Specifications
- Description:Polypropylene Buchner Funnel
- Cat. no.:470174-670
- Supplier No.:57105
Specifications
About this item
Useable With or Without Filter Paper.
- Uses 7.0 cm diameter filter paper
Two-piece HDPE funnel with sealed-in, polyethylene filter plate of coarse porosity offers good retention and fast flow under vacuum.
Customers Who Bought This Also Bought