Polyester Seine
Catalog #: 470018-572
Supplier: Peco Sales
Specifications
- Description:Polyester Seine
- Cat. no.:470018-572
- Mesh Size:1/4"
- Length:10'
- Width:4'
- Supplier No.:77699
Specifications
About this item
Great for Stream Sampling
- Large Area Screen for Sampling
- Collect More Fish and Specimens
It measures 10' long, 4' feet high, and is constructed of 1/4" polyester mesh. It comes with floats and leads, but not poles.
Customers Who Bought This Also Bought