Kemtec® Introduction To Plant Cloning
Catalog #: 470111-206
Supplier: Kemtec
Specifications
- Description:Plant Cloning Kit I
- Cat. no.:470111-206
- Supplier No.:2-504
Specifications
About this item
Learn to clone plants.
- Practice sterile technique
- No hood or incubator required
Cloning plant tissue makes an excellent introduction to biotechnology and sterile technique as plant propagation by cloning becomes increasingly important in food and flower production. Students germinate carrot seeds in a sterile environment to provide plant material from which to establish a culture, use sterile technique to grow seedlings and establish a callus culture, and induce the development of multiple shoots (plantlets) from callus tissue. The process takes approximately four months.
Ordering information: This item contains a coupon for perishable materials that must be redeemed from the manufacturer before use. Please allow sufficient time to account for redemption. These materials are shipped from the supplier via second day delivery on Mondays, Tuesdays, or Wednesdays only and must be received promptly.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought