Photographic Fog
Catalog #: 470024-590
Supplier: FORENSICS SOURCE
Specifications
- Description:Photographic Fog
- Cat. no.:470024-590
- Supplier No.:PF-1
Specifications
About this item
View the Trajectory of a Laser
For easier viewing and photographing of a laser’s trajectory, a few short bursts of photographic fog pointed at a laser beam make its entire length visible. 8 oz. can.
Customers Who Bought This Also Bought